Assay Details
| Gene Symbol: UBE2J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194458.1 | NP_919440.1 | 2 - 3 | 453 | 140 | |
| XM_011540616.2 | XP_011538918.1 | 2 - 3 | 442 | 262 | ||
| GenBank mRNA | BG112933.1 | - | 448 | 140 | ||
| BM926567.1 | - | 2 - 3 | 450 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_194458.1, XM_011540616.2 transcripts.
Set Membership: |
|

UBE2J2
Genomic Map