Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160407.1 | NP_001153879.1 | 1 - 2 | 181 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001306.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17017452.hg.1 | ENST00000394933.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017452.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017452.hg.1 | TC17001306.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180205.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180205.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180205.hg.1 | TC1700010209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180205.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180205.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000292090.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000580518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | ENST00000581236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010209.hg.1 | NM_138463.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180204.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180204.hg.1 | NM_001160407.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180204.hg.1 | TC1700010209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180204.hg.1 | ENST00000394933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180204.hg.1 | NM_001160407.1 | Refseq |

TLCD1
Genomic Map