Assay Details
| Gene Symbol: FAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014613.2 | NP_055428.1 | 5 - 6 | 536 | 108 | |
| XM_011534475.2 | XP_011532777.1 | 4 - 5 | 423 | 108 | ||
| GenBank mRNA | AB020694.1 | - | 5 - 6 | 478 | 108 | |
| AB088120.1 | - | 5 - 6 | 489 | 108 | ||
| AK304346.1 | - | 5 - 6 | 506 | 108 | ||
| BC001069.1 | - | 5 - 6 | 514 | 2961467 | 108 | |
| BC006145.1 | - | 5 - 6 | 495 | 3942891 | 108 | |
| BC014001.2 | - | 5 - 6 | 495 | 3936113 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001000.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
lipid particle organization
negative regulation of catalytic activity
protein binding
ubiquitin protein ligase binding
lipase binding
ubiquitin binding
lipase inhibitor activity
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
lipid particle organization
negative regulation of catalytic activity
protein binding
ubiquitin protein ligase binding
lipase binding
ubiquitin binding
lipase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05014376.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014376.hg.1 | ENST00000540174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014376.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014376.hg.1 | TC05001000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014378.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014378.hg.1 | ENST00000540174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014378.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014378.hg.1 | TC05001000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014375.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014375.hg.1 | ENST00000540174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014375.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014375.hg.1 | TC05001000.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167107.hg.1 | TC0500009591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500167104.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167104.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167104.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167104.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167104.hg.1 | TC0500009591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167105.hg.1 | TC0500009591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000261942.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000504983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000506061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000510730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | ENST00000513627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009591.hg.1 | NM_014613.2 | Refseq |

FAF2
Genomic Map