Assay Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003226.1 | 6 - 7 | 772 | 90 | ||
| NR_003228.1 | 6 - 7 | 738 | 90 | |||
| GenBank mRNA | AF329691.1 | - | 5 - 6 | 523 | 90 | |
| AK091293.1 | - | 6 - 7 | 735 | 90 | ||
| BC021245.2 | - | 4 - 5 | 345 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CENPB DNA-binding domain containing 1 | CENPBD1 | -11.7kb | Anti-sense | Chr. 16 - 89969775 - 89972832 on NCBI Build GRCh38 | |
| Downstream | dysbindin domain containing 1 | DBNDD1 | 15.9kb | Anti-sense | Chr. 16 - 90004865 - 90020131 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000710.hg
Additional Information:
This assay is in the 5'UTR region of the AK091293.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000355531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000388970.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000418696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000421780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000423742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000427658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000429663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000431757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000436447.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000437774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000450026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000454997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000457926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000458301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | ENST00000557444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003226.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003227.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008908.hg.1 | NR_003228.1 | Refseq |

AFG3L1P
Genomic Map