| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000303824.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000340726.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000388740.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000388741.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000388742.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000388743.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000388744.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
ENST00000419131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
NM_000503.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
NM_172058.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
NM_172059.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
NM_172060.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016955.hg.1 |
TC08001316.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000303824.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000340726.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388740.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388741.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388742.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388743.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000419131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000465115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000493349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000496494.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_000503.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_001288574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_001288575.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172058.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172060.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000303824.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000340726.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388740.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388741.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388742.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000388743.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000419131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000465115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000493349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
ENST00000496494.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_000503.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_001288574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_001288575.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172058.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172059.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
NM_172060.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173141.hg.1 |
TC0800010774.hg.1 |
Clariom D TC |