Assay Details
| Gene Symbol: STRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003162.3 | NP_003153.2 | 9 - 10 | 1191 | 97 | |
| XM_005264519.4 | XP_005264576.1 | 8 - 9 | 1215 | 97 | ||
| XM_011533073.2 | XP_011531375.1 | 10 - 11 | 1413 | 97 | ||
| GenBank mRNA | AJ223814.1 | - | 9 - 10 | 1191 | 97 | |
| AK026934.1 | - | 6 - 7 | 762 | 97 | ||
| AK307836.1 | - | 5 - 6 | 858 | 97 | ||
| BC106878.2 | - | 10 - 11 | 1108 | 97 | ||
| BC106879.2 | - | 9 - 10 | 1043 | 40027865 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001736.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
locomotory behavior
negative regulation of cell proliferation
Wnt signaling pathway
dendrite development
bicellular tight junction assembly
protein binding
calmodulin binding
estrogen receptor binding
protein complex binding
protein phosphatase 2A binding
armadillo repeat domain binding
locomotory behavior
negative regulation of cell proliferation
Wnt signaling pathway
dendrite development
bicellular tight junction assembly
protein binding
calmodulin binding
estrogen receptor binding
protein complex binding
protein phosphatase 2A binding
armadillo repeat domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027905.hg.1 | ENST00000263918.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027905.hg.1 | ENST00000379213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027905.hg.1 | ENST00000538092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027905.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027905.hg.1 | TC02001736.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189337.hg.1 | TC0200012252.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000263918.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000379213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000495595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | ENST00000624376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012252.hg.1 | NM_003162.3 | Refseq |

STRN
Genomic Map