| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015406.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015406.hg.1 |
ENST00000373657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015406.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015406.hg.1 |
TC0X001137.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015407.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015407.hg.1 |
ENST00000373657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015407.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015407.hg.1 |
TC0X001137.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015404.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015404.hg.1 |
ENST00000373657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015404.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X015404.hg.1 |
TC0X001137.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168393.hg.1 |
TC0X00010025.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168391.hg.1 |
TC0X00010025.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010025.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
ENST00000334463.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
ENST00000373657.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
NM_017669.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00168389.hg.1 |
TC0X00010025.hg.1 |
Clariom D TC |