| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
ENST00000334318.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
ENST00000343327.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
ENST00000374255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
ENST00000374257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
ENST00000458258.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_001163788.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_001163790.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_001244896.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_001244897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_001244898.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
NM_005156.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018483.hg.1 |
TC09001488.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000334318.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000343327.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000374255.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000374257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000458258.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001163788.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001163790.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244896.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244898.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_005156.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000334318.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000343327.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000374255.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000374257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
ENST00000458258.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001163788.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001163790.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244896.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_001244898.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
NM_005156.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176376.hg.1 |
TC0900011217.hg.1 |
Clariom D TC |