Assay Details
| Gene Symbol: CELSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001408.2 | NP_001399.1 | 16 - 17 | 6167 | 62 | |
| XM_005270580.4 | XP_005270637.1 | 16 - 17 | 6481 | 62 | ||
| GenBank mRNA | AF234887.1 | - | 16 - 17 | 6168 | 62 | |
| AK309093.1 | - | 7 - 8 | 966 | 62 | ||
| D87469.2 | - | 16 - 17 | 5899 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000941.hg.1
Set Membership: |
|

CELSR2
Genomic Map