| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014864.hg.1 |
ENST00000271332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014864.hg.1 |
ENST00000498157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014864.hg.1 |
NM_001408.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014864.hg.1 |
TC01000941.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000271332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000489018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000498157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
NM_001408.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000271332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000489018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
ENST00000498157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
NM_001408.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166696.hg.1 |
TC0100009345.hg.1 |
Clariom D TC |