Assay Details
| Gene Symbol: NUSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243142.1 | NP_001230071.1 | 5 - 6 | 803 | 167 | |
| NM_001243143.1 | NP_001230072.1 | 5 - 6 | 761 | 167 | ||
| NM_001243144.1 | NP_001230073.1 | 4 - 5 | 737 | 167 | ||
| NM_001301136.1 | NP_001288065.1 | 5 - 6 | 803 | 167 | ||
| NM_016359.4 | NP_057443.2 | 5 - 6 | 806 | 167 | ||
| NM_018454.7 | NP_060924.4 | 5 - 6 | 806 | 167 | ||
| XM_005254428.2 | XP_005254485.1 | 5 - 6 | 678 | 167 | ||
| XM_005254430.4 | XP_005254487.1 | 5 - 6 | 600 | 167 | ||
| XM_005254431.2 | XP_005254488.1 | 5 - 6 | 678 | 167 | ||
| XM_006720559.2 | XP_006720622.1 | 5 - 6 | 678 | 167 | ||
| XM_006720560.3 | XP_006720623.1 | 5 - 6 | 774 | 167 | ||
| XM_006720561.2 | XP_006720624.1 | 5 - 6 | 592 | 167 | ||
| XM_006720562.2 | XP_006720625.1 | 5 - 6 | 678 | 167 | ||
| XM_006720563.2 | XP_006720626.1 | 5 - 6 | 660 | 167 | ||
| XM_017022294.1 | XP_016877783.1 | 5 - 6 | 600 | 167 | ||
| XM_017022295.1 | XP_016877784.1 | 5 - 6 | 1470 | 167 | ||
| GenBank mRNA | AF090915.1 | - | 5 - 6 | 610 | 166 | |
| AF290612.1 | - | 5 - 6 | 610 | 167 | ||
| AF305711.1 | - | 5 - 6 | 542 | 167 | ||
| AK023483.1 | - | 5 - 6 | 616 | 167 | ||
| AK225703.1 | - | 5 - 6 | 617 | 167 | ||
| AK290879.1 | - | 5 - 6 | 630 | 167 | ||
| AK293168.1 | - | 4 - 5 | 561 | 167 | ||
| BC001308.1 | - | 5 - 6 | 609 | 3453354 | 167 | |
| BC010838.1 | - | 3 - 4 | 350 | 167 | ||
| BC012887.1 | - | 5 - 6 | 584 | 3882061 | 167 | |
| BC024772.1 | - | 5 - 6 | 611 | 4342499 | 167 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF090915.1, BC012887.1, XM_017022295.1 transcripts.
Set Membership: |
|

NUSAP1
Genomic Map