Assay Details
| Gene Symbol: IQSEC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170738.1 | NP_001164209.1 | 9 - 10 | 2712 | 71 | |
| NM_015232.1 | NP_056047.1 | 8 - 9 | 2215 | 71 | ||
| XM_011520958.2 | XP_011519260.1 | 9 - 10 | 2800 | 71 | ||
| XM_011520959.2 | XP_011519261.1 | 9 - 10 | 2800 | 71 | ||
| XM_011520960.1 | XP_011519262.1 | 9 - 10 | 2799 | 71 | ||
| XM_011520961.1 | XP_011519263.1 | 8 - 9 | 2215 | 71 | ||
| XM_017019310.1 | XP_016874799.1 | 9 - 10 | 2800 | 71 | ||
| XM_017019311.1 | XP_016874800.1 | 9 - 10 | 2800 | 71 | ||
| GenBank mRNA | AB029033.1 | - | 6 - 7 | 1387 | 71 | |
| AK091953.1 | - | 8 - 9 | 2215 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000003.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000544511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | ENST00000544511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | ENST00000326261.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | ENST00000538872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12000041.hg.1 | TC12000003.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144067.hg.1 | TC1200006438.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000382841.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000537151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000538872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000540907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000544511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | ENST00000544511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_001170738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006438.hg.1 | NM_015232.1 | Refseq |

IQSEC3
Genomic Map