| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000348370.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000557998.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000559209.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000561186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270528.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270529.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270530.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_017610.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
TC1500010747.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000348370.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000557998.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000559209.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
ENST00000561186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270528.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270529.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_001270530.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500182543.hg.1 |
NM_017610.7 |
Refseq |