Assay Details
| Gene Symbol: STARD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020151.3 | NP_064536.2 | 1 - 2 | 683 | 68 | |
| GenBank mRNA | AF270647.1 | - | 1 - 2 | 610 | 68 | |
| AK054615.1 | - | 1 - 2 | 49 | 68 | ||
| AK291190.1 | - | 1 - 2 | 65 | 68 | ||
| AL542720.3 | - | 1 - 2 | 685 | 68 | ||
| BC003028.1 | - | 1 - 2 | 666 | 68 | ||
| BC007894.2 | - | 2 - 3 | 441 | 4134933 | 68 | |
| BC008894.2 | - | 1 - 2 | 666 | 3833465 | 68 | |
| BC009998.2 | - | 2 - 3 | 441 | 4128869 | 68 | |
| BC012774.2 | - | 1 - 2 | 633 | 3842611 | 68 | |
| BC012793.2 | - | 1 - 2 | 647 | 4080200 | 68 | |
| BC013279.2 | - | 1 - 2 | 666 | 3830320 | 68 | |
| BC014076.2 | - | 1 - 2 | 643 | 4127007 | 68 | |
| BC014274.2 | - | 1 - 2 | 662 | 3677198 | 68 | |
| BC032106.1 | - | 1 - 2 | 657 | 4643387 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002104.hg
Additional Information:
This assay is in the 5'UTR region of the AK054615.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034011.hg.1 | ENST00000337288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034011.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034011.hg.1 | TC02002104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034008.hg.1 | ENST00000337288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034008.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034008.hg.1 | TC02002104.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198823.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198827.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198827.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198827.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198827.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198827.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198828.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198825.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198825.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198825.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198825.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198825.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198826.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198826.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198826.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198826.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198826.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198822.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000462501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000479456.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | ENST00000495687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013515.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | ENST00000488084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198824.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | ENST00000337288.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | ENST00000443962.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | NM_020151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200198829.hg.1 | TC0200013515.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 113628 | - | - |

STARD7
Genomic Map