Assay Details
| Gene Symbol: RANBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300865.1 | NP_001287794.1 | 3 - 4 | 342 | 107 | |
| NM_007320.2 | NP_015559.2 | 3 - 4 | 342 | 107 | ||
| XM_006722929.1 | XP_006722992.1 | 3 - 4 | 184 | 118 | ||
| XM_011528391.1 | XP_011526693.1 | 3 - 4 | 292 | 107 | ||
| XM_011528393.1 | XP_011526695.1 | 3 - 4 | 137 | 118 | ||
| GenBank mRNA | AK222813.1 | - | 3 - 4 | 126 | 107 | |
| AK301942.1 | - | 3 - 4 | 126 | 107 | ||
| AK314343.1 | - | 3 - 4 | 173 | 107 | ||
| BC004349.1 | - | 3 - 4 | 124 | 3350317 | 107 | |
| BM786367.1 | - | 3 - 4 | 342 | 107 | ||
| DQ234346.1 | - | 3 - 4 | 115 | 107 | ||
| Y08697.1 | - | 2 - 3 | 107 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006722929.1, XM_011528391.1, XM_011528393.1 transcripts.
Set Membership: |
|

RANBP3
Genomic Map