Assay Details
| Gene Symbol: MIPOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020999.1 | XP_016876488.1 | 8 - 9 | 981 | 133 | |
| XM_017021000.1 | XP_016876489.1 | 4 - 5 | 465 | 133 | ||
| XM_017021001.1 | XP_016876490.1 | 5 - 6 | 604 | 133 | ||
| XM_017021008.1 | XP_016876497.1 | 4 - 5 | 463 | 133 | ||
| XM_017021009.1 | XP_016876498.1 | 5 - 6 | 576 | 133 | ||
| XM_017021012.1 | XP_016876501.1 | 5 - 6 | 576 | 133 | ||
| GenBank mRNA | BC036523.1 | - | 4 - 5 | 388 | 4828885 | 133 |
| BX647795.1 | - | 4 - 5 | 359 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC036523.1,XM_017020999.1,XM_017021000.1,XM_017021001.1,XM_017021008.1,XM_017021009.1,XM_017021012.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MIPOL1
Genomic Map