Assay Details
| Gene Symbol: SPTLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018327.2 | NP_060797.2 | 2 - 3 | 596 | 102 | |
| XM_011529279.1 | XP_011527581.1 | 2 - 3 | 364 | 102 | ||
| XM_017027922.1 | XP_016883411.1 | 2 - 3 | 364 | 102 | ||
| GenBank mRNA | AK300184.1 | - | 2 - 3 | 402 | 102 | |
| BC150644.1 | - | 2 - 3 | 555 | 9057014 | 102 | |
| CF994231.1 | - | 2 - 3 | 370 | 102 | ||
| CX870585.1 | - | 1 - 2 | 89 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000092.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000485952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | ENST00000485952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001263.hg.1 | ENST00000378194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001263.hg.1 | ENST00000399002.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001263.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001263.hg.1 | TC20000092.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | TC2000006736.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146206.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000399002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000431275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000434210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000450297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000476791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000485952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | ENST00000485952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006736.hg.1 | NM_018327.3 | Refseq |

SPTLC3
Genomic Map