Assay Details
| Gene Symbol: DSC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001941.4 | NP_001932.2 | 6 - 7 | 931 | 135 | |
| NM_024423.3 | NP_077741.2 | 6 - 7 | 931 | 135 | ||
| GenBank mRNA | D17427.1 | - | 6 - 7 | 843 | 135 | |
| X83929.1 | - | 7 - 8 | 797 | 135 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000435.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004422.hg.1 | ENST00000360428.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004422.hg.1 | ENST00000434452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004422.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004422.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004422.hg.1 | TC18000435.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | ENST00000584980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008385.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | NM_024423.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | TC1800008385.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | ENST00000360428.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | ENST00000434452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | NM_001941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800157117.hg.1 | NM_024423.3 | Refseq |

DSC3
Genomic Map