Assay Details
| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269926.4 | XP_005269983.1 | 1 - 2 | 380 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005269926.4 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
Wnt signaling pathway
cell migration
regulation of cell morphogenesis
positive regulation of Wnt signaling pathway
protein K29-linked deubiquitination
protein K63-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
protein K33-linked deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
zinc ion binding
K63-linked polyubiquitin binding
cytoskeleton organization
Wnt signaling pathway
cell migration
regulation of cell morphogenesis
positive regulation of Wnt signaling pathway
protein K29-linked deubiquitination
protein K63-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
protein K33-linked deubiquitination
thiol-dependent ubiquitin-specific protease activity
protein binding
zinc ion binding
K63-linked polyubiquitin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000359653.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000471421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | ENST00000508096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009221.hg.1 | NM_017580.2 | Refseq |

ZRANB1
Genomic Map