Assay Details
| Gene Symbol: CCDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025004.2 | NP_079280.2 | 10 - 11 | 2394 | 93 | |
| XM_017018346.1 | XP_016873835.1 | 11 - 12 | 2514 | 93 | ||
| XM_017018347.1 | XP_016873836.1 | 12 - 13 | 2424 | 93 | ||
| XM_017018349.1 | XP_016873838.1 | 10 - 11 | 3655 | 93 | ||
| XM_017018350.1 | XP_016873839.1 | 9 - 10 | 2271 | 93 | ||
| XM_017018352.1 | XP_016873841.1 | 10 - 11 | 2164 | 93 | ||
| GenBank mRNA | AK023277.1 | - | 10 - 11 | 2394 | 93 | |
| BC018540.1 | - | 10 - 11 | 2308 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001150.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014486.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014486.hg.1 | ENST00000529051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014486.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014486.hg.1 | TC11001150.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170533.hg.1 | TC1100009407.hg.1 | Clariom D TC |

CCDC15
Genomic Map