Assay Details
| Gene Symbol: CCDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025004.2 | NP_079280.2 | 8 - 9 | 2163 | 98 | |
| GenBank mRNA | AK023277.1 | - | 8 - 9 | 2163 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001150.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11014484.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014484.hg.1 | ENST00000529051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014484.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014484.hg.1 | TC11001150.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170531.hg.1 | TC1100009407.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000344762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000524471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000529051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | ENST00000530061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009407.hg.1 | NM_025004.2 | Refseq |

CCDC15
Genomic Map