Assay Details
| Gene Symbol: COX6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144613.4 | NP_653214.2 | 2 - 3 | 298 | 61 | |
| GenBank mRNA | AK057427.1 | - | 2 - 3 | 195 | 61 | |
| AY152398.1 | - | 2 - 3 | 204 | 61 | ||
| BC026123.2 | - | 2 - 3 | 299 | 4827471 | 61 | |
| BC064548.1 | - | 2 - 3 | 180 | 61 | ||
| BC100899.2 | - | 1 - 2 | 135 | 40003591 | 61 | |
| BC100900.1 | - | 1 - 2 | 135 | 40003592 | 61 | |
| BC100901.1 | - | 1 - 2 | 135 | 40003593 | 61 | |
| BC100902.1 | - | 1 - 2 | 135 | 40003594 | 61 | |
| BG717038.1 | - | 2 - 3 | 303 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001869.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025439.hg.1 | ENST00000326529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025439.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025439.hg.1 | TC19001869.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | TC1900012051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208346.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |

COX6B2
Genomic Map