Assay Details
| Gene Symbol: CNKSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173515.2 | NP_775786.2 | 7 - 8 | 1235 | 75 | |
| XM_011535485.2 | XP_011533787.1 | 7 - 8 | 3718 | 75 | ||
| GenBank mRNA | AK055911.1 | - | 7 - 8 | 1275 | 75 | |
| AK309680.1 | - | 7 - 8 | 841 | 75 | ||
| AY328891.1 | - | 7 - 8 | 1235 | 75 | ||
| AY328892.1 | - | 7 - 8 | 1235 | 75 | ||
| AY328893.1 | - | 7 - 8 | 1235 | 75 | ||
| AY328894.1 | - | 4 - 5 | 518 | 75 | ||
| BC060761.1 | - | 7 - 8 | 1157 | 5299236 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004140.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06028274.hg.1 | ENST00000367213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028274.hg.1 | ENST00000433165.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028274.hg.1 | ENST00000479339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028274.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028274.hg.1 | TC06004140.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000424998.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014361.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000433165.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000454664.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000479339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | ENST00000607772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | NM_173515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202956.hg.1 | TC0600014361.hg.1 | Clariom D TC |

CNKSR3
Genomic Map