Assay Details
| Gene Symbol: ALDH1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270364.1 | NP_001257293.1 | 13 - 14 | 1877 | 57 | |
| NM_001270365.1 | NP_001257294.1 | 11 - 12 | 1676 | 57 | ||
| NM_012190.3 | NP_036322.2 | 13 - 14 | 1979 | 57 | ||
| XM_006713481.2 | XP_006713544.1 | 13 - 14 | 1718 | 57 | ||
| XM_011512355.1 | XP_011510657.1 | 13 - 14 | 1711 | 57 | ||
| GenBank mRNA | AB209299.1 | - | 13 - 14 | 1786 | 57 | |
| AF052732.1 | - | 13 - 14 | 1739 | 57 | ||
| AK222535.1 | - | 13 - 14 | 1761 | 57 | ||
| AK222694.1 | - | 13 - 14 | 1761 | 57 | ||
| AK294392.1 | - | 11 - 12 | 1460 | 57 | ||
| AK308112.1 | - | 13 - 14 | 1718 | 57 | ||
| CR749807.1 | - | 13 - 14 | 1877 | 57 | ||
| KU178541.1 | - | 13 - 14 | 1813 | 57 | ||
| KU178542.1 | - | 12 - 13 | 1629 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ALDH1L1
Genomic Map