Assay Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004867.4 | NP_004858.1 | 2 - 3 | 580 | 88 | |
| GenBank mRNA | AF038953.1 | - | 2 - 3 | 382 | 88 | |
| AK223327.1 | - | 2 - 3 | 359 | 88 | ||
| AK313154.1 | - | 2 - 3 | 538 | 88 | ||
| AY359051.1 | - | 2 - 3 | 321 | 88 | ||
| BC040437.1 | - | 2 - 3 | 335 | 5748519 | 88 | |
| CR456875.1 | - | 2 - 3 | 243 | 88 | ||
More Information
Set Membership: |
|

ITM2A
Genomic Map