Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001425.2 | NP_001416.1 | 1 - 2 | 243 | 69 | |
| GenBank mRNA | AL541088.3 | - | 1 - 2 | 243 | 69 | |
| BC009718.1 | - | 1 - 2 | 50 | 3891050 | 69 | |
| U52101.1 | - | 1 - 2 | 38 | 69 | ||
| U87947.1 | - | 1 - 2 | 230 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000699.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC009718.1,NM_001425.2,U52101.1,U87947.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19009665.hg.1 | ENST00000270221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009665.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009665.hg.1 | TC19000699.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009667.hg.1 | ENST00000270221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009667.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009667.hg.1 | TC19000699.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009666.hg.1 | ENST00000270221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009666.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19009666.hg.1 | TC19000699.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | TC1900011774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201829.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000593437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011774.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | TC1900011774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201820.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | NM_001425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | TC1900011774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000270221.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000594198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000596315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000597279.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000597529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | ENST00000599255.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201825.hg.1 | NM_001425.2 | Refseq |

EMP3
Genomic Map