| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017568.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017568.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017568.hg.1 |
TC04001285.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011016.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174905.hg.1 |
TC0400011016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
ENST00000514391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174903.hg.1 |
TC0400011016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174904.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174904.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174904.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174904.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174904.hg.1 |
TC0400011016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174906.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174906.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174906.hg.1 |
ENST00000513150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174906.hg.1 |
NR_026769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174906.hg.1 |
TC0400011016.hg.1 |
Clariom D TC |