Assay Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005245069.2 | XP_005245126.1 | 1 - 2 | 63 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005245069.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
transmembrane transport
divalent metal ion transport
cellular response to magnesium ion
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
substrate-specific transporter activity
magnesium:sodium antiporter activity
divalent inorganic cation transmembrane transporter activity
cellular magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
transmembrane transport
divalent metal ion transport
cellular response to magnesium ion
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
substrate-specific transporter activity
magnesium:sodium antiporter activity
divalent inorganic cation transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |

SLC41A1
Genomic Map