| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
ENST00000484228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017079.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
ENST00000367137.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
ENST00000468057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
NM_173854.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229430.hg.1 |
TC0100017079.hg.1 |
Clariom D TC |