| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000302946.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000359345.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372611.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000372613.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
ENST00000439569.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001168247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_001300859.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013853.hg.1 |
NM_144990.3 |
Refseq |