Assay Details
| Gene Symbol: GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020474.3 | NP_065207.2 | 9 - 10 | 1492 | 149 | |
| XM_005258239.3 | XP_005258296.1 | 10 - 11 | 1560 | 149 | ||
| XM_017025692.1 | XP_016881181.1 | 10 - 11 | 1739 | 149 | ||
| GenBank mRNA | AJ505952.1 | - | 2 - 3 | 231 | 149 | |
| BC038440.1 | - | 10 - 11 | 1557 | 4478643 | 149 | |
| X85018.1 | - | 9 - 10 | 1398 | 149 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000145.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001655.hg.1 | ENST00000269195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001655.hg.1 | ENST00000537549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001655.hg.1 | ENST00000537748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001655.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001655.hg.1 | TC18000145.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | NM_020474.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | TC1800007080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000269195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000589189.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | ENST00000590654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148568.hg.1 | NM_020474.3 | Refseq |

GALNT1
Genomic Map