Assay Details
| Gene Symbol: C5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306149.1 | NP_001293078.1 | 1 - 2 | 331 | 87 | |
| NM_001306150.1 | NP_001293079.1 | 1 - 2 | 331 | 87 | ||
| NM_178569.3 | NP_848664.1 | 1 - 2 | 331 | 87 | ||
| XM_005248256.3 | XP_005248313.1 | 1 - 2 | 522 | 87 | ||
| XM_005248257.3 | XP_005248314.1 | 1 - 2 | 522 | 87 | ||
| XM_011513975.2 | XP_011512277.1 | 1 - 2 | 522 | 87 | ||
| XM_011513977.2 | XP_011512279.1 | 1 - 2 | 522 | 87 | ||
| GenBank mRNA | AY249325.1 | - | 1 - 2 | 314 | 87 | |
| BC101608.1 | - | 1 - 2 | 234 | 8069114 | 87 | |
| BC101634.1 | - | 1 - 2 | 234 | 8069140 | 87 | |
| BC143639.1 | - | 1 - 2 | 234 | 9052153 | 87 | |
| CX754076.1 | - | 1 - 2 | 216 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000026.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | ENST00000334000.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | ENST00000397835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | ENST00000505778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | ENST00000515640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000395.hg.1 | TC05000026.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000457752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001294337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006558.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145014.hg.1 | TC0500006558.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000334000.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000397835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000503940.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000505106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000505778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | ENST00000515640.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_001306149.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_001306150.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NM_178569.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NR_122125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | NR_122126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145015.hg.1 | TC0500006558.hg.1 | Clariom D TC |

C5orf38
Genomic Map