| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000361099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000392322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000392323.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000409465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000540176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
ENST00000544783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
NM_007315.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
NM_139266.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02042032.hg.1 |
TC02002624.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000361099.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000392322.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000392323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000409465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000452281.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000540176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
NM_007315.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
NM_139266.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000361099.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000392322.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000392323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000409465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000452281.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
ENST00000540176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
NM_007315.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
NM_139266.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211660.hg.1 |
TC0200015242.hg.1 |
Clariom D TC |