Assay Details
| Gene Symbol: PAX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511793.1 | XP_011510095.1 | 8 - 9 | 1095 | 119 | |
| XM_011511794.1 | XP_011510096.1 | 8 - 9 | 1064 | 119 | ||
| GenBank mRNA | KU178405.1 | - | 7 - 8 | 897 | 119 | |
More Information
Set Membership: |
|

PAX8
Genomic Map