Assay Details
| Gene Symbol: GCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006836.1 | NP_006827.1 | 36 - 37 | 4557 | 63 | |
| GenBank mRNA | BC153881.1 | - | 36 - 37 | 4557 | 8860118 | 63 |
| D86973.2 | - | 36 - 37 | 4557 | 63 | ||
| U77700.1 | - | 14 - 15 | 2022 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002034.hg.1
Set Membership: |
|

GCN1
Genomic Map