Assay Details
| Gene Symbol: GCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006836.1 | NP_006827.1 | 40 - 41 | 5167 | 114 | |
| GenBank mRNA | BC153881.1 | - | 40 - 41 | 5167 | 8860118 | 114 |
| D86973.2 | - | 40 - 41 | 5167 | 114 | ||
| U77700.1 | - | 18 - 19 | 2632 | 114 | ||
| U88836.1 | - | 1 - 2 | 125 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002034.hg.1
Set Membership: |
|

GCN1
Genomic Map