Assay Details
| Gene Symbol: GCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006836.1 | NP_006827.1 | 48 - 49 | 6408 | 58 | |
| GenBank mRNA | BC040172.1 | - | 4 - 5 | 569 | 58 | |
| BC153881.1 | - | 48 - 49 | 6408 | 8860118 | 58 | |
| D86973.2 | - | 48 - 49 | 6408 | 58 | ||
| U77700.1 | - | 26 - 27 | 3873 | 58 | ||
| U88836.1 | - | 9 - 10 | 1366 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002034.hg.1
Set Membership: |
|

GCN1
Genomic Map