| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
ENST00000451092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
ENST00000543224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023389.hg.1 |
TC03001364.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
ENST00000451092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
ENST00000543224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023393.hg.1 |
TC03001364.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000330022.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000431180.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000445387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000330022.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000431180.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000445387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300180077.hg.1 |
TC0300010968.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000330022.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000431180.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000445387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000330022.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000409792.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000431180.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000445387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
ENST00000492397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
NM_014159.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300180080.hg.1 |
TC0300010968.hg.1 |
Clariom D TC |