| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
ENST00000253861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
ENST00000460346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
ENST00000539845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
ENST00000545148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
ENST00000546185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
NM_021807.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013031.hg.1 |
TC07000829.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000253861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000460346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000469115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000479839.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000482089.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000483800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000486013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000486409.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000489931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
NM_021807.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000253861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000460346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000469115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000479839.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000482089.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000483800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000486013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000486409.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
ENST00000489931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
NM_021807.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165485.hg.1 |
TC0700009211.hg.1 |
Clariom D TC |