Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017741.3 | NP_060211.3 | 2 - 3 | 423 | 132 | |
| XM_006713966.3 | XP_006714029.1 | 3 - 4 | 582 | 132 | ||
| XM_017008327.1 | XP_016863816.1 | 1 - 2 | 1876 | 132 | ||
| GenBank mRNA | AK056116.1 | - | 2 - 3 | 423 | 132 | |
| BC050697.1 | - | 1 - 2 | 84 | 6044417 | 132 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001069.hg
Additional Information:
This assay is in the 5'UTR region of the AK056116.1,BC050697.1,NM_017741.3,XM_006713966.3,XM_017008327.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014721.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014721.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014721.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014721.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014722.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014722.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014722.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014722.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014724.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014724.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014724.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014724.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014724.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014723.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014723.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014723.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014723.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014723.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014730.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014730.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014730.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014730.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014730.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014727.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014727.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014727.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014727.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014720.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014720.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014720.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014720.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014726.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014726.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014726.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014726.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014729.hg.1 | ENST00000382247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014729.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014729.hg.1 | ENST00000536863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014729.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014729.hg.1 | TC04001069.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400169694.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169694.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169694.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169694.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169694.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169697.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010167.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169704.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169700.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169700.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169700.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169700.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169700.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169696.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169696.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169696.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169696.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169696.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169698.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169701.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169701.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169701.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169701.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169701.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000507731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | ENST00000507768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169703.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169695.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169695.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169695.hg.1 | ENST00000382247.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169695.hg.1 | NM_017741.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169695.hg.1 | TC0400010167.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198670 | - | - |

DCAF16
Genomic Map