Assay Details
| Gene Symbol: RNGTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286426.1 | NP_001273355.1 | 11 - 12 | 1488 | 109 | |
| NM_001286428.1 | NP_001273357.1 | 10 - 11 | 1378 | 109 | ||
| GenBank mRNA | AB009023.1 | - | 11 - 12 | 1268 | 109 | |
| AB009024.1 | - | 11 - 12 | 1268 | 109 | ||
| AK299777.1 | - | 10 - 11 | 1368 | 109 | ||
More Information
Set Membership: |
|

RNGTT
Genomic Map