Assay Details
| Gene Symbol: FXR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004860.3 | NP_004851.2 | 11 - 12 | 1473 | 86 | |
| GenBank mRNA | AK226082.1 | - | 3 - 4 | 284 | 86 | |
| AK313836.1 | - | 11 - 12 | 1158 | 86 | ||
| BC020090.1 | - | 11 - 12 | 1427 | 4385430 | 86 | |
| BC051907.1 | - | 11 - 12 | 1304 | 6149976 | 86 | |
| BC067272.1 | - | 11 - 12 | 1418 | 6204341 | 86 | |
| U31501.1 | - | 11 - 12 | 1335 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001092.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014668.hg.1 | ENST00000250113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014668.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014668.hg.1 | TC17001092.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000571597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000573957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000574490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | ENST00000576693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009649.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174912.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174912.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174912.hg.1 | TC1700009649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700174912.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174912.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | NM_004860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | TC1700009649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | ENST00000250113.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | ENST00000571079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174913.hg.1 | NM_004860.3 | Refseq |

FXR2
Genomic Map