Assay Details
| Gene Symbol: SH3GL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301108.1 | NP_001288037.1 | 2 - 3 | 416 | 129 | |
| NM_001324185.1 | NP_001311114.1 | 3 - 4 | 567 | 129 | ||
| NM_001324187.1 | NP_001311116.1 | 2 - 3 | 354 | 129 | ||
| NR_136711.1 | 3 - 4 | 567 | 129 | |||
| NR_136713.1 | 2 - 3 | 416 | 129 | |||
| NR_136714.1 | 2 - 3 | 416 | 129 | |||
| XM_017022486.1 | XP_016877975.1 | 2 - 3 | 521 | 129 | ||
| GenBank mRNA | AF036270.1 | - | 2 - 3 | 113 | 129 | |
| AK307442.1 | - | 2 - 3 | 268 | 129 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | basonuclin 1 | BNC1 |
BNC, BSN1, HsT19447 |
-274.5kb | Anti-sense | Chr. 15 - 83255884 - 83284716 on NCBI Build GRCh38 |
| Downstream | ADAMTS like 3 | ADAMTSL3 | ADAMTSL-3 | 88.9kb | Sense | Chr. 15 - 83654086 - 84039842 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF036270.1, NM_001301108.1, NM_001324185.1, NM_001324187.1, XM_017022486.1 transcripts.
Set Membership: |
|

SH3GL3
Genomic Map