Assay Details
| Gene Symbol: CASP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001228.4 | NP_001219.2 | 4 - 5 | 707 | 77 | |
| GenBank mRNA | U60520.1 | - | 4 - 5 | 660 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

CASP8
Genomic Map