Assay Details
| Gene Symbol: BRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011515184.2 | XP_011513486.1 | 2 - 3 | 166 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011515184.2 transcripts.
Set Membership: |
|

BRAT1
Genomic Map