Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TVP23C-CDRT4 readthrough | TVP23C-CDRT4 | FAM18B2-CDRT4 | Anti-sense | Chr. 17 - 15436015 - 15563631 on NCBI Build GRCh38 | |
| Downstream | CMT1A duplicated region transcript 1 | CDRT1 |
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2 |
58.1kb | Anti-sense | Chr. 17 - 15565482 - 15619704 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map