Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928106 | LOC101928106 | -49.7kb | Anti-sense | Chr. 6 - 10465999 - 10481946 on NCBI Build GRCh38 | |
| Downstream | chromosome 6 open reading frame 52 | C6orf52 | 137.6kb | Anti-sense | Chr. 6 - 10671418 - 10694797 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map