| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004858.hg.1 |
ENST00000374388.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004858.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004858.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004858.hg.1 |
TC0X000376.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00151642.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151642.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151642.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151642.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151642.hg.1 |
TC0X00007537.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000374403.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
ENST00000485406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007537.hg.1 |
NM_012310.4 |
Refseq |