| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
ENST00000219476.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
ENST00000350773.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
ENST00000353929.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
ENST00000401874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
ENST00000439673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001211.hg.1 |
TC16000059.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000219476.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000350773.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000353929.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000382538.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000401874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
ENST00000439673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001209.hg.1 |
TC16000059.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000219476.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000350773.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000353929.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000382538.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000401874.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
ENST00000439673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16001212.hg.1 |
TC16000059.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
TC1600006587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146179.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
TC1600006587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146178.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000382538.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
TC1600006587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000382538.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
ENST00000568454.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146180.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000382538.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_001114382.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
TC1600006587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000219476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000350773.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000382538.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000401874.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000439117.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000439673.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
ENST00000461648.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_000548.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_001077183.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600146176.hg.1 |
NM_001114382.2 |
Refseq |