Assay Details
| Gene Symbol: LOC107987373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006724896.2 | XP_006724959.1 | 2 - 3 | 230 | 74 | |
| XM_017030111.1 | XP_016885600.1 | 2 - 3 | 230 | 74 | ||
| Gene Symbol: MRPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021134.3 | NP_066957.3 | 2 - 3 | 230 | 74 | |
| XM_006718271.2 | XP_006718334.1 | 2 - 3 | 230 | 74 | ||
| XM_011520273.1 | XP_011518575.1 | 2 - 3 | 230 | 74 | ||
| XM_011520275.2 | XP_011518577.1 | 2 - 3 | 230 | 74 | ||
| XM_011520276.2 | XP_011518578.1 | 2 - 3 | 230 | 74 | ||
| GenBank mRNA | AK313805.1 | - | 2 - 3 | 170 | 74 | |
| BC027710.2 | - | 2 - 3 | 145 | 4298000 | 74 | |
| U26596.1 | - | 2 - 3 | 193 | 74 | ||
| Z49254.1 | - | 2 - 3 | 193 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000055.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | ENST00000381514.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | ENST00000381519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | ENST00000397294.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | ENST00000397297.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | ENST00000397298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001103.hg.1 | TC11000055.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000484918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | ENST00000486931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006547.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000381514.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000381519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397294.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397297.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000397298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000429295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000462288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | ENST00000466346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | NM_021134.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145757.hg.1 | TC1100006547.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC11_KI270721v1_random00006432.hg.1 | ENST00000612848.1 | Ensembl |

LOC107987373
Genomic Map